missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GCV Protein
A cDNA sequence encoding the GCV was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11945-1000mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
GCV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Greater than 99.0%. |
1 g | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |