missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL11 / Eotaxin Protein
A DNA sequence encoding the Rat CCL11 / Eotaxin, was expressed in the hosts and tags indicated.
105.00€ - 2370.00€
Especificaciones
Número de acceso | NP_062078.1 |
---|---|
ID de gen (Entrez) | 29397 |
Nombre | CCL11 / Eotaxin Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15953408
|
enQuireBio™
QP10296-ec-5ug |
5 μg | N/A | ||||||
15933408
|
enQuireBio™
QP10296-ec-20ug |
20 μg | N/A | ||||||
15923408
|
enQuireBio™
QP10296-ec-100ug |
100 μg | N/A | ||||||
15943408
|
enQuireBio™
QP10296-ec-250ug |
250 μg | N/A | ||||||
Especificaciones
NP_062078.1 | |
CCL11 / Eotaxin Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
HPGSIPTSCC FTMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTKLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP | |
Greater than 96% as determined by SDS-PAGE and HPLC. |
29397 | |
Research Use Only | |
CCL11 | |
The biological activity determined by a chemotaxis bioassay using purified eosinophils is in a concentration range of 0.1-1.0 ug/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |