missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 7 Protein
A cDNA sequence encoding the IL 7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10720-2ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
25647 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 was determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is less than 0.2ng/ml, corresponding to a specific activity of >5,000,000IU/mg. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
IL 7 Protein | |
Research Use Only | |
Il7 | |
Recombinant Protein | |
E. coli | |
DCHIKDKDGK AFGSVLMISI NQLDKMTGTD SDCPNNEPNF FKKHLCDDTK EAAFLNRAAR KLRQFLKMNI SEEFNDHLLR VSDGTQTLVN CTSKEEKTIK EQKKNDPCFL KRLLREIKTC WNKILKGSI | |
Greater than 98.0% as determined by SDS-PAGE. |