Learn More
enQuireBio™ Recombinant Rat MCP-3 / CCL7 Protein
A DNA sequence encoding the Rat MCP-3 / CCL7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10295-ec-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001007613.1 | |
MCP-3 / CCL7 Protein | |
Research Use Only | |
CCL7 | |
The biologically active determined by a chemotaxis bioassay using human monocytes is in a concentration range of 10-100 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
287561 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
QPDGTNSSTC CYVKKQKIPK RNLKSYRKIT SSRCPWEAVI FKTKKGMEVC AEAHQKWVEE AIAYLDMKTS TPKP | |
Greater than 95% as determined by SDS-PAGE and HPLC. |