missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Protein-lysine 6-oxidase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Protein-lysine 6-oxidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6314-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_058757.1 | |
Protein-lysine 6-oxidase Protein | |
Research Use Only | |
Lox | |
Recombinant Protein | |
E. coli | |
DDPYNPYKYS DDNPYYNYYD TYERPRSGSR HRPGYGTGYF QYGLPDLVPD PYYIQASTYV QKMSMYNLRC AAEENCLASS AYRADVRDYD HRVLLRFPQR VKNQGTSDFL PSRPRYSWEW HSCHQHYHSM DEFSHYDLLD ASTQRRVAEG HKASFCLEDT SCDYGYHRRF ACTAHTQGLS PGCYDTYAAD IDCQWIDITD VQPGNYILKV SVNPSYLVPE SDYSNNVVRC EIRYTGHHAY ASGCTISPY | |
Greater than 90% as determined by SDS-PAGE. |
24914 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |