Learn More
R&D Systems™ Recombinant SARS-CoV-2 NSP16/NSP10-His Complex
Marca: R&D Systems™ 10634-CV-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
YP_009725311.1 (NSP16) and YP_009725306.1 (NSP10) | |
100 μg | |
SARS-CoV-2 |
no conjugado | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl, TCEP and Glycerol. |