missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant SARSCoV-2 B.1.1.529 RBD Alexa Fluor™ 488 Protein

Código de producto. 18305337
Change view
Click to view available options
Cantidad:
20 μg
Tamaño de la unidad:
1 set
Código de producto. Cantidad unitSize
18305337 20 μg 1 set
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18305337

Marca: R&D Systems™ AFG11056020

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Omicron Variant His-tag

Measured by flow cytometry for its ability to bind HEK293 human embryonic kidney cells transfected with human ACE-2 at 0.250-1.00 μg/mL (100 μL/well). Please Note: Optimal dilutions should be determined by each laboratory for each application.
TRUSTED_SUSTAINABILITY

Specifications

Número de acceso YP_009724390.1
Para utilizar con (aplicación) Bioactivity
Formulación Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Peso molecular M.W. (Observed): 33-39 kDa, under reducing conditions., M.W. (Theoretical): 26 kDa
Cantidad 20 μg
Fuente Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike RBD protein Arg319-Phe541 (Gly339Asp, Ser371Leu, Ser373Pro, Ser375Phe, Lys417Asn, Asn440Lys, Gly446Ser, Ser477Asn, Thr478Lys, Glu484Ala, Gln493Arg, Gly496Ser, Gln498Arg, Asn501Tyr, Tyr505His), with a C-terminal 6-His tag
Requisitos de almacenamiento Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Concentración de endotoxinas <1.0 EU per 1 μg of the protein by the LAL method.
Conjugado Alexa Fluor 488
Grado de pureza o calidad >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Expresión Spike RBD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.