missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zebrafish Metallothionein-2 Protein
A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) Metallothionein-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9826-ye-200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001124525.2 | |
Metallothionein-2 Protein | |
Research Use Only | |
mt2 | |
Recombinant Protein | |
Yeast | |
MDPCECAKTG TCNCGATCKC TNCQCTTCKK SCCSCCPSGC SKCASGCVCK GNSCGSSCCQ | |
Greater than 90% as determined by SDS-PAGE. |
100174951 | |
200 μg | |
Not determined. | |
Full Length | |
Zebrafish | |
His | |
Tris-based buffer, 50% glycerol |