Learn More
Gilac™ Rectangular HACCP Container with Lid
Rectangular HACCP Container with Lid is a food container for large storage volumes; lid can support the weight of heavy loads.
Marca: Gilac™ G119611
Detalles adicionales : Peso : 1.51000kg
Descripción
- HACCP rectangular food container.
- For large storage volumes, lid can support the weight of heavy loads.
- Multipurpose container.
- Rewritable identification cartridge for inventory tracking.
- No gaps nor overlaps.
- Machine washable.
- Recessed handles for better handling.
- Stackable without lid.
- Compatible with all GILAC 4 and 6 wheel dollies.
Especificaciones
Storage Tote | |
Green | |
4 gal. |
HDPE | |
Rectangular | |
15 L |