Learn More
Thermo Scientific™ Remel™ Letheen Broth (Dehydrated)
The Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.
Marca: Thermo Scientific™ R453712
201.67 EUR hasta el 2024-12-31
Use el código promocional "23263" para aplicar la oferta.
Detalles adicionales : Peso : 0.01000kg
Descripción
Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.
Especificaciones
Referencia industrial: AOAC. | |
Polvo | |
500 g |
Caldo Letheen | |
Medios de cultivo deshidratados |
REF R07700, REF R112420, REF R112423, and REF R117259 not intended for IVD use.