missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Rhesus macaque GITR / TNFRSF18 Protein, Fc Tag
423.38€ - 2661.24€
Especificaciones
Peso molecular | 41.3 kDa |
---|---|
Fuente | Rhesus macaque GITR, Fc Tag (GIR-C5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # F7DDV8). |
Inmunógeno | Gln 26 - Glu 161 |
Requisitos de almacenamiento | -20°C |
Alias de gen | AITR,GITR,TNFRSF18,CD357 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15806847
|
ACROBiosystems
GIR-C5255-1MG |
1 mg |
2661.24€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15896837
|
ACROBiosystems
GIR-C5255-100UG |
100 μg |
423.38€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Glucocorticoid-induced TNFR-related protein (GITR) is also known as Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), activation-inducible TNFR family receptor (AITR), CD antigen CD357, which is a member of the tumor necrosis factor receptor (TNF-R) superfamily. GITR is receptor for TNFSF18, which seems to be involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. GITR also mediated NF-kappa-B activation via the TRAF2/NIK pathway.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.3 kDa. As a result of glycosylation, the protein migrates as 48 kDa under reducing (R) condition, and 85 kDa under non-reducing (NR) condition (SDS-PAGE).Especificaciones
41.3 kDa | |
Gln 26 - Glu 161 | |
AITR,GITR,TNFRSF18,CD357 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Rhesus macaque GITR, Fc Tag (GIR-C5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # F7DDV8). | |
-20°C | |
GITR | |
Rhesus Macaque | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |