missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ribosomal protein L4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11302-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L4E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Especificaciones
| ribosomal protein L4 | |
| Polyclonal | |
| Unconjugated | |
| RPL4 | |
| 60S ribosomal protein L1, 60S ribosomal protein L4, ribosomal protein L4, RPL1, RPL4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6124, 64302, 67891 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P36578, P50878, Q9D8E6 | |
| RPL4 | |
| ribosomal protein L4 Fusion Protein Ag1828 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido