missing translation for 'onlineSavingsMsg'
Learn More

Rislenemdaz, MedChemExpress

Código de producto. 16464088
Encompass_Preferred
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
50 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
5 miligramos
50 miligramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16464088

Marca: Medchem Express HY106441A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Rislenemdaz (CERC-301) is an orally bioavailable and selective N-methyl-D-aspartate (NMDA) receptor subunit 2B (GluN2B) antagonist with Ki and IC 50 of 8.1 nM and 3.6 nM, respectively.

Rislenemdaz (CERC-301) is an orally bioavailable and selective N-methyl-D-aspartate (NMDA) receptor subunit 2B (GluN2B) antagonist with Ki and IC 50 of 8.1 nM and 3.6 nM, respectively.

Specifications

Nombre del producto químico o material Rislenemdaz
Forma física Solid
CAS 808732-98-1
Color Blanco
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C19H23FN4O2
Para utilizar con (aplicación) Neuroscience-Neurodegeneration
Sinónimo MK-0657 CERC-301
Información de solubilidad DMSO : 100 mg/mL (279.01 mM; Need ultrasonic)
SMILES O=C(N1C[C@@H](F)[C@@H](CNC2=NC=CC=N2)CC1)OCC3=CC=C(C)C=C3
Peso molecular (g/mol) 358.41
Formula Weight (peso de la fórmula) 358.41
Porcentaje de pureza 97.5%
Grado Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.