Learn More
RNF139 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA518148
338.68 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene is a multi-membrane spanning protein containing a RING-H2 finger. This protein is located in the endoplasmic reticulum, and has been shown to possess ubiquitin ligase activity. This gene was found to be interrupted by a ttranslocation in a family with hereditary renal and non-medulary thyroid cancer. Studies of the Drosophila counterpart suggested that this protein may interact with tumor suppressor protein VHL, as well as with COPS5/JAB1, a protein responsible for the degradation of tumor suppressor CDKN1B/P27KIP.
Especificaciones
RNF139 | |
Polyclonal | |
Unconjugated | |
RNF139 | |
4930555P18Rik; E3 ubiquitin-protein ligase RNF139; HRCA1; MGC31961; multiple membrane spanning receptor TRC8; patched related protein translocated in renal cancer; RCA1; RING finger protein 139; RING finger protein 139; E3 ubiquitin-protein ligase RNF139; RING-type E3 ubiquitin transferase RNF139; RNF139; translocation in renal carcinoma on chromosome 8 protein; TRC8 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
11236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8WU17 | |
RNF139 | |
Synthetic peptide sequence (HTGAAAEEFNDDT) corresponding to the C-terminus amino acids of RNF139 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.