Learn More
Invitrogen™ RNF43 Recombinant Rabbit Monoclonal Antibody (077)

Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA530821
Descripción
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human RNF43.
E3 ubiquitin-protein ligase that acts as a negative regulator of the Wnt signaling pathway by mediating the ubiquitination, endocytosis and subsequent degradation of Wnt receptor complex components Frizzled. Acts on both canonical and non-canonical Wnt signaling pathway. Acts as a tumor suppressor in the intestinal stem cell zone by inhibiting the Wnt signaling pathway, thereby resticting the size of the intestinal stem cell zone.
Especificaciones
| RNF43 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q68DV7 | |
| RNF43 | |
| Recombinant Human RNF43 Protein (Met1-Tyr197). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 077 | |
| Unconjugated | |
| RNF43 | |
| 4732452J19Rik; E3 ubiquitin-protein ligase RNF43; RGD1305204; RING finger protein 43; RING-type E3 ubiquitin transferase RNF43; RNF124; Rnf43; URCC | |
| Rabbit | |
| Protein A | |
| RUO | |
| 54894 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.