Learn More
RNF8 Mouse, anti-Human, Clone: 3G3H5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68521-1-IG-20UL
Descripción
The RNF8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RNF8. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The RNF8 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
RNF8 | |
3G3H5 | |
FLJ12013, KIAA0646, ring finger protein 8, RNF8 | |
20 μL | |
9025 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |
Western Blot, ELISA | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Rabbit, Pig | |
150 kDa | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.