Learn More
STEMCELL Technologies RoboSep™ Human Pan-B Cell Enrichment Kit
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Marca: STEMCELL Technologies 19554RF
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The EasySep™ Human Pan-B Cell Enrichment Kit is designed to isolate B cells including plasma cells from normal fresh or previously frozen peripheral blood mononuclear cells by negative selection
- Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD36, CD42b, CD56, CD66b, CD123, glycophorin A and dextran-coated magnetic particles
- The labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Especificaciones
B Cells, Plasma | |
Cell Isolation | |
1 x 109 cells |