Learn More
STEMCELL Technologies RoboSep™ Human Pan-B Cell Enrichment Kit
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Marca:  STEMCELL Technologies 19554RF
Encompass
Código de producto. 17187921
  • 1906.50€ / 1 set
Fecha estimada de envío: 23-10-2024
para ver el stock
Explore más ofertas especiales
Añadir a la cesta
Descripción
Descripción
  • The EasySep™ Human Pan-B Cell Enrichment Kit is designed to isolate B cells including plasma cells from normal fresh or previously frozen peripheral blood mononuclear cells by negative selection
  • Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD36, CD42b, CD56, CD66b, CD123, glycophorin A and dextran-coated magnetic particles
  • The labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
Especificaciones
Especificaciones
B Cells, Plasma
Cell Isolation
1 x 109 cells