Learn More
Thermo Scientific™ Selenite Lactose Broth
Reduce preparation time when enriching for Salmonella species from fecal and food samples with ready-to-use prepared media tubes.
57.40€
Especificaciones
Color Marfil claro transparente
Tipo de producto Caldo
Ver más especificaciones
Productos 1
Código de producto Marca Descripción Cantidad Volumen de llenado Precio Cantidad y disponibilidad  
Código de producto Marca Descripción Cantidad Volumen de llenado Precio Cantidad y disponibilidad  
13225799
Ver documentos Detalles de la promoción
Thermo Scientific™
TV5005I
Caldo de lactosa de selenito 50/paq. 8 mL
57.40€
paquete de 50
En stock
para ver el stock
Añadir a la cesta
Descripción
Descripción
Enrich for Salmonella species from fecal and food samples with Thermo Scientific™ Selenite Lactose Broth in convenient prepared media tubes.
Selenite Lactose Broth is used for enrichment culture of salmonellae from feces, food products, water and other materials.
Sodium biselenite acts as the selective agent, while lactose is added as a fermentable carbohydrate to prevent a rise in pH value during incubation because any increase in pH will reduce the selective activity of selenite.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificaciones
Especificaciones
Marfil claro transparente
Caldo
Videos
Seguridad y manipulación
Seguridad y manipulación
missing translation for 'shelfLife' : 26 semanas
SDS
Documentos
Documentos
Product Certifications
For In Vitro Diagnostic Use.