Learn More
Sensititre™ EUGNF
Código de producto. 17928630
Click to view available options
Tipo de caldo:
MHB (n.° de referencia T3462)
Tamaño de la unidad:
cada uno
Código de producto. 17928630
Marca: Thermo Scientific Oxoid EUGNF
Perform susceptibility testing of non-fastidious Gram-Negative isolates on a single, convenient plate which meets EUCAST breakpoints.
Perform susceptibility testing of non-fastidious Gram-Negative species isolates on a single platform with Thermo Scientific™ Sensititre™ Gram-Negative EUGNF AST Plate.
The convenient standard AST plate meets EUCAST breakpoints for relevant antimicrobials on the plate including Colistin, Ceftazidime/Avibactam and Ceftolozane/Tazobactam .
Achieve reliable, consistent antimicrobial susceptibility results for non-fastidious Gram-Negative isolates with the Sensititre Gram-Negative EUGNF AST Plate. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.
  • Monitor susceptibility among Gram-negative isolates with full-range, accurate MIC values.
  • For ultimate flexibility automated or manual read options are available
  • Eliminate inventory control concerns with extended shelf life (18 to 24 months) and room temperature storage.
The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
  • Scalable: flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Flexible: over 240 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made: design your own AST panel to meet specific formulary requirements and prescription protocols and to clinicians that are based on real-time trends and local epidemiology
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Especificaciones
Patrón McFarland 0,5 Demineralized Water (part no. T3339)
Agentes antimicrobianos Diverso
Tipo de caldo MHB (n.° de referencia T3462)
Descripción Sensititre Gram Negative EUGNF AST Plate
Código de antibiótico Various
Tipo de producto AST Plate
Clase de organismo diana Non-fastidious, Gram-negative Organisms
Tipo de prueba Minimum Inhibitory Concentration (MIC)
Condiciones de incubación 35°C; O2
Tiempo de incubación 18-24 hrs
Aísla por placa 1
Tipo de empaquetado Box
Cantidad 10/Box
Opciones de lectura Autoread or manual
Volumen de transferencia 10μL
Volumen por pocillo 50 μl