Learn More
Serpin A5/Protein C Inhibitor Antibody (110), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP2-89406B
Descripción
Serpin A5/Protein C Inhibitor Monoclonal antibody specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for ELISA
Especificaciones
Serpin A5/Protein C Inhibitor | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
110 | |
ELISA | |
Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A5/Protein C Inhibitor (NP_000615.3; Met 1-Pro 406). | |
0.1 mL | |
Cancer | |
5104 | |
Store at 4°C in the dark. | |
IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.