Learn More
SHOWA 8110 Cut-Resistant Knit Gloves
Click to view available options
Tamaño:
Pequeño
M
Descripción
- The gloves are designed to fit the hand naturally, reducing fatigue during prolonged use.
- They offer optimal dexterity and breathability, keeping hands cool and comfortable even during intense physical activity.
- The gloves are ideal for industries that require reliable cut protection, such as construction and manufacturing.
- With exceptional durability, superior protection, and unparalleled comfort, the SHOWA 8110 gloves are the go-to choice for hand protection needs.
Especificaciones
Especificaciones
Coating Material | Uncoated |
Color | Gris |
Hand | Ambidiestro |
Latex Free | Sí |
Material | Glass Fiber, Polyethylene |
Tipo de producto | Cut-resistant Gloves |
Tamaño | Pequeño |
Norma EN388 | EN388:2X43E |
Sexo | Unisex |
Estilo de puño de los guantes | Knit |