Learn More
SHOWA S-TEX KV3 Cut-Resistant Knit Gloves
Click to view available options
Tamaño:
Pequeño
M
L
XL
Descripción
- The glove has a natural rubber palm dip that ensures a superior grip in both dry and wet conditions.
- For maximum cut resistance, the KV3 glove is made with a combination of stainless steel liner, Kevlar fibers, and patented Hagane Coil technology.
- The KV3 glove is designed to be form-fitting and breathable, ensuring maximum comfort and performance during use.
Especificaciones
Especificaciones
Estructura | Seamless Knit |
Coating Material | Látex |
Color | Negro, Yellow |
Hand | Específico |
Latex Free | No |
Material | Aramid Knit, Polyester, Stainless Steel Fiber |
Tipo de producto | Cut-resistant Gloves |
Tamaño | Pequeño |
Sexo | Unisex |
Estilo de puño de los guantes | Knit |