missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHPRH Mouse anti-Human, Clone: OTI4A3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF501448
Detalles adicionales : Peso : 0.05000kg
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.Especificaciones
SHPRH | |
Monoclonal | |
Unconjugated | |
SHPRH | |
bA545I5.2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
257218 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q149N8 | |
SHPRH | |
Human recombit protein fragment corresponding to amino acids 1-100 and 1500 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |