missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A4 Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543571
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Carnitine (b-hydroxy-g-trimethylaminobutyrate) is a small, highly polar compound that aids in the b-oxidation of long-chain fatty acids. Organic cation/carnitine transporters (OCTN) assist in the elimination of cationic compounds, including xenobiotics, and transport carnitine for reabsorption in the kidney. Similar to organic cation transporters (OCT), OCTN proteins localize to the plasma membrane of epithelial cells. OCTN1 is expressed in kidney, trachea, bone marrow and fetal liver. OCTN2 is abundantly expressed in kidney, skeletal muscle, placenta and heart. OCTN3 is strongly expressed in testis and weakly expressed in kidney. The gene encoding human OCTN1 maps to chromosome 5 and the gene encoding human OCTN2 maps to chromosome 5q31. Mutations in the gene encoding OCTN2 leads to systemic carnitine deficiency (SCD), an autosomal recessive disorder characterized by cardiomyopathy, skeletal myopathy, lethargy, hypoglycemia and hyperammonemia.Especificaciones
SLC22A4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9H015 | |
SLC22A4 | |
Leu42-Lys141 (Accession Q9H015), with N-terminal His Tag | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Western Blot | |
C1 | |
Unconjugated | |
SLC22A4 | |
ergothioneine transporter; ET transporter; ETT; integral membrane transport protein; LSTP-Like 2; Octn1; organic cation transporter 1; organic cation transporter OCTN1; organic cation/carnitine transporter 1; SLC22A4; solute carrier family (organic cation transporter), member 4; solute carrier family 22 (organic cation transporter), member 4; solute carrier family 22 (organic cation/ergothioneine transporter), member 4; solute carrier family 22 (organic cation/zwitterion transporter), member 4; solute carrier family 22 member 4; UT2H | |
Mouse | |
Protein A/G | |
RUO | |
6583 | |
-20°C | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido