Learn More
SLC27A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA567531
Product Code. 15872714
  • 472.00€ / 100 microlitros
Explore more special offers
This item is not returnable. View return policy
Description
Description
This gene encodes a member of a family of fatty acid transport proteins, which are involved in translocation of long-chain fatty acids cross the plasma membrane. This protein is expressed at high levels on the apical side of mature enterocytes in the small intestine, and appears to be the principal fatty acid transporter in enterocytes. Clinical studies suggest this gene as a candidate gene for the insulin resistance syndrome. Mutations in this gene have been associated with ichthyosis prematurity syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC27A4
Polyclonal
Unconjugated
SLC27A4
ACSVL4; BB144259; Fatp4; FATP-4; fatty acid transport protein 4; IPS; Long-chain fatty acid transport protein 4; SLC27A4; solute carrier family 27 (fatty acid transporter), member 4; solute carrier family 27 member 4
Rabbit
affinity chromatography
RUO
10999
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6P1M0
SLC27A4
Fusion protein of human SLC27A4
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions