missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA539553
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Slc30A8 is a zinc efflux transporter involved in the accumulation of zinc in intracellular vesicles and is expressed at a high level in the pancreas, particularly in islets of Langerhans, and co-localizes with insulin in the secretory pathway granules of the insulin-secreting INS-1 cells. Allelic variants of Slc30A8 have been identified as major genetic risk factors for the development of Type 2 diabetes, but no genetic association has been found with Type 1 diabetes. Slc30A8 expression in b-cells was found to be influenced by cytokine expression, particularly IFN-gamma and IL-1 beta.Especificaciones
SLC30A8 | |
Polyclonal | |
Unconjugated | |
SLC30A8 | |
ZNT8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
169026 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8IWU4 | |
SLC30A8 | |
A synthetic peptide derived from the internal region of human SLC30A8 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |