Learn More
Invitrogen™ SOD2 (MnSOD) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5116561
Descripción
Positive Samples: 293T, HepG2, A-549, Mouse liver, Mouse brain, Mouse heart, Rat liver, Rat brain, Rat heart Immunogen sequence: KHSLPDLPYD YGALEPHINA QIMQLHHSKH HAAYVNNLNV TEEKYQEALA KGDVTAQIAL QPALKFNGGG HINHSIFWTN LSPNGGGEPK GELLEAIKRD FGSFDKFKEK LTAASVGVQG SGWGWLGFNK ERGHLQIAAC PNQDPLQGTT GLIPLLGIDV WEHAYYLQYK NVRPDYLKAI WNVINWENVT ERYMACKK.
Superoxide dismutase (SOD) is an antioxidant enzyme involved in the defense system against reactive oxygen species (ROS). SOD catalyzes the dismutation reaction of superoxide radical anion (O2) to hydrogen peroxide, which is then catalyzed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each of Cu and Zn ion per subunit. The manganese containing 80 kDa tetrameric enzyme SOD2, is located in the mitochondrial matrix in close proximity to a primary endogenous source of superoxide, the mitochondrial respiratory chain. SOD3 is a heparin-binding multimer of disulfide-linked dimers, primarily expressed in human lungs, vessel walls and airways. SOD4 is a copper chaperone for superoxide dismutase (CCS), which specifically delivers Cu to copper/zinc superoxide dismutase. CCS may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor.
Especificaciones
| SOD2 (MnSOD) | |
| Polyclonal | |
| Unconjugated | |
| SOD2 | |
| I79_015272; indophenoloxidase B; IPO B; IPOB; IPO-B; manganese SOD; manganese superoxide dismutase; manganese-containing superoxide dismutase; mangano-superoxide dismutase; manganous superoxide dismutase; mitochondrial superoxide dismutase 2; Mn superoxide dismutase; MNSOD; Mn-SOD; MVCD6; similar to manganous superoxide dismutase; Sod2; Sod-2; sod2.L; sodMt; Superoxide dimutase 2, mitochondrial; superoxide dismutase; superoxide dismutase (Mn type); superoxide dismutase [Mn], mitochondrial; superoxide dismutase 2; superoxide dismutase 2 L homeolog; superoxide dismutase 2, mitochondrial; superoxide dismutase 2, mitochondrial L homeolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20656, 24787, 6648 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.57 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P04179, P07895, P09671 | |
| SOD2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-222 of human SOD2 (NP_0006272). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.