missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SON Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA565107
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: TCMVSETPAMS AEPTVLASEP PVMSETAETF DSMRASGHVA SEVSTSLLVP AVTTPVLAES ILEPPAMAAP ESSAMAVLES SAVTVLESST VTVL Highest antigen sequence identity to the following orthologs - mouse 56%, rat 54%.
The protein encoded by this gene binds to a specific DNA sequence upstream of the upstream regulatory sequence of the core promoter and second enhancer of human hepatitis B virus (HBV). Through this binding, it represses HBV core promoter activity, transcription of HBV genes, and production of HBV virions. The protein shows sequence similarities with other DNA-binding structural proteins such as gallin, oncoproteins of the MYC family, and the oncoprotein MOS. It may also be involved in protecting cells from apoptosis and in pre-mRNA splicing. Several transcript variants encoding different isoforms have been described for this gene, but the full-length nature of only two of them has been determined. [provided by RefSeq].Especificaciones
SON | |
Polyclonal | |
Unconjugated | |
SON | |
2900011L12Rik; AA409051; AU067731; BASS1; Bax antagonist selected in Saccharomyces 1; C21orf50; C81487; DBP5; DBP-5; HSPC310; HSPC312; KIAA1019; mKIAA1019; negative regulatory element-binding protein; NRE-binding protein; NREBP; Protein DBP-5; Protein SON; SON; Son cell proliferation protein; Son DNA binding protein; SON DNA-binding protein; SON3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6651 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P18583 | |
SON | |
Recombinant Human SON | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only