Learn More
SORT/TPL/G Calibration And Maintenance Buffers, Gel Electodes, WTW™
Catalog Number 10599052
Click to view available options
Cantidad:
250 x 2 mL
Calibration and maintenance are complete assortments for the calibration and maintenance of electrodes. The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCI as a storage or refill solution.
Cantidad 250 x 2 mL