missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEN Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB100-58799R
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SPEN Polyclonal antibody specifically detects SPEN in Human, Mouse samples. It is validated for Western BlotEspecificaciones
SPEN | |
Polyclonal | |
Western Blot | |
HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila) | |
The immunogen recognized by this antibody maps to a region between residue 3375 and 3425 of human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator) using the numbering given in entry NP_055816.2 (GeneID 23013). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23013 | |
Store at 4C in the dark. | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido