Learn More
Cole-Parmer™ Hydraulic Laboratory Pellet Press Accessories, Die Set Pellet
Optimize performance of your Cole-Parmer™ hydraulic laboratory pellet press with this Die Set Pellet.
Marca: Cole-Parmer™ 3613
Detalles adicionales : Peso : 0.45400kg
Descripción
- Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more.
- Ensure top results from your Cole-Parmer™ hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components.
- The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.
Especificaciones
Die Set Pellet | |
13 mm | |
For IR Analysis | |
2.2 lb. |
0.5 in. | |
To Form 13 mm KBR Disks | |
Evacuable die body, base, plunger, two 13 mm polished steel pellets, o-ring vacuum seals and knock out ring for sample disk extraction | |
1 kg |