Learn More
Brady™ Static Dissipative Thermal Transfer Printable Labels
Printable, static dissipative polyester labels for Thermal Transfer Printers.
Marca: Brady™ 054552
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Material is UL and CSA approved with its respective ribbon
- Ideal for Circuit Board Labeling
- For smooth surfaces
Especificaciones
Not Autoclavable | |
Permanent Acrylic | |
Gloss | |
Thermal Transfer | |
Circuit Board Labeling | |
1 | |
Square | |
0.09 mm | |
12.7 mm |
THT-99-473-10 | |
White | |
Static-dissipative Polyester | |
No estéril | |
Impresoras de las series BBP™81, BradyPrinter i5100, i7100, PR Plus e IP™, impresoras THT con núcleo de 76 mm | |
5000 Labels | |
De -40 °C a +120 °C | |
15.2 mm | |
12.7 mm |