missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Stella Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA523449
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b) and 94% homologous in chimp.
DPPa3 (stella-related protein) is a stem cell marker protein which is restricted to pluripotent human embryonic stem cells, premeiotic germ lineage cells and testicular germ cell tumor cells. Otherwise known as 'developmental pluripotency-associated protein 3,' it is expressed in primordial germ cells, Oocytes, preimplantation embryos and pluripotent cells. This protein has a SAP-like domain and a splicing factor motif-like structure, suggesting possible roles in chromosomal organization or RNA processing.
Especificaciones
| Stella | |
| Polyclonal | |
| Unconjugated | |
| DPPA3 | |
| 2410075G02Rik; Cap1p; Compaction-associated protein 1; Crg1; developmental pluripotency associated 3; developmental pluripotency-associated 3; developmental pluripotency-associated protein 3; Dppa3; PCG7; Pgc7; primordial germ cell 7; primordial germ cell protein 7; Stella; STELLAR; Stella-related protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 359787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q6W0C5 | |
| DPPA3 | |
| A portion of amino acids 100-159 of human STELLAR (isoform CRA_b). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido