Learn More
Streptozotocin, 97+%, Thermo Scientific™
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Streptozotocin acts as a bactericidal by alkylating DNA and inhibiting DNA synthesis
• Streptozotocin enters cells through glucose transporter 2
Application
• Streptozotocin induces diabetes in experimental animals and can cause acute liver injury
• It is also useful for research on neuroendocrine tumors and carcinoma cells
Especificaciones
Especificaciones
Nombre del producto químico o material | Streptozotocin |
Color | White |
Cantidad | 500mg |
Sensibilidad | Hygroscopic |
Beilstein | 2060675 |
Índice Merck | 14,8832 |
Información de solubilidad | Soluble in water or 100% ethanol |
Formula Weight (peso de la fórmula) | 265.22 |
Porcentaje de pureza | ≥97% |
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.