Descripción
- Use with clean, dry air.
- Ideal for gas mixing, valve operations, and calibration stands.
- These pressure regulators are designed for precise regulation of air pressure in pipes, vessels, and process lines.
- Regulators maintain constant pressure by providing high accuracy, precision control, and maximum stability under the most adverse conditions.
- Regulators feature high-accuracy pressure regulation up to ±0.1%, a servo-operated control mechanism for pressure stability, and start-up stability that returns to output setting after a long down-time.
- Locking nut prevents inadvertant adjustment of pressure.
- Unit can be in-line or panel mounted.
Especificaciones
Especificaciones
| Descripción | Regulador subminiatura de 0 a 0,344 bar (de 0 a 5 psig); NPT (hembra) de 0,157 cm (0,062 pulg) |
| Tipo | Regulador de gas |
| Para utilizar con (equipo) | Cilindro |
| Material | Latón |
| Diámetro del calibre (métrico) | 29,46 mm |