missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SuperNuclease Recombinant Rabbit Monoclonal Antibody (026), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543451
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: SuperNuclease (Recombinant Serratia marcescens Endonuclease)
SuperNuclease is a genetically-engineered endonuclease. It is a nonspecific nuclease with high activity, capable of completely digesting RNA and DNA (single-stranded, double-stranded, linear, circular and supercoiled forms, that no fewer than five phosphate residues) into 5'-monophosphate-terminated oligonucleotides of 3-5 bases in length. SuperNuclease can be removed by various purification methods.Especificaciones
SuperNuclease | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG |
ELISA | |
026 | |
Unconjugated | |
DNA endonuclease; RNA endonuclease | |
SuperNuclease (Recombinant Serratia marcescens Endonuclease) | |
50 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |