missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV39H1 Monoclonal Antibody (44.1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA125505
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: HeLa nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.Especificaciones
SUV39H1 | |
Monoclonal | |
2.4 mg/mL | |
0.01M PBS with 15mM sodium azide; pH 7.4 | |
O43463, O54864 | |
SUV39H1 | |
Recombinant fusion protein, MBP-SUV39H1. | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
44.1 | |
Unconjugated | |
SUV39H1 | |
AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; KMT1A; Lysine N-methyltransferase 1A; MG44; mIS6; Position-effect variegation 3-9 homolog; RGD1565028; Su(var)3-9 homolog 1; suppressor of variegation 3-9 homolog 1; suppressor of variegation 3-9 homolog 1 (Drosophila); SUV39H; Suv39h histone methyltransferase; SUV39H1 | |
Mouse | |
purified | |
RUO | |
20937, 302553, 6839 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |