missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBXA2R Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | TBXA2R |
---|---|
Concentración | 0.27 mg/mL |
Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Code produit | Marque | Cantidad | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Cantidad | Prix | Quantité et disponibilité | |||||
16825445
|
Proteintech
27159-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16815445
|
Proteintech
27159-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.Spécification
TBXA2R | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P21731, P30987 | |
21390, 6915 | |
TBXA2R Fusion Protein Ag25670 | |
Primary | |
-20°C | |
TBXA2R |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Prostanoid TP receptor, TBXA2R, thromboxane A2 receptor, TXA2 R | |
TBXA2R | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |