missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10779-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).Especificaciones
TCEB2 | |
Polyclonal | |
Unconjugated | |
ELOB | |
ELOB, Elongin 18 kDa subunit, Elongin B, SIII, SIII p18, TCEB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67673, 6923, 81807 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62869, P62870, Q15370 | |
Elob | |
TCEB2 Fusion Protein Ag1228 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |