missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR Cβ1 Mouse anti-Human, BV750, Clone: JOVI.1, BD Biosciences™
Description
TCR Cβ1 Monoclonal antibody specifically detects TCR Cβ1 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antígeno | TCR Cβ1 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | JOVI.1 |
| Conjugado | Brilliant Violet 750 |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Alias de gen | T-cell receptor beta locus; TCRB; TRB; TRB@; TRCB1 |
| Especie del huésped | Mouse |
| Inmunógeno | Lymphoid cells from Mouse transgenic for Human HA1.7 cell TCRbeta |
| Cantidad | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?