Learn More
Thermo Scientific Chemicals Teriflunomida
Click to view available options
Cantidad:
250 mg
1 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Nombre del producto químico o material | Teriflunomida |
Cantidad | 1 g |
Punto de fusión | de 229,0 °C |
Formula Weight (peso de la fórmula) | 270,21 |
Clave InChI | UTNUDOFZCWSZMS-UHFFFAOYSA-N |
SMILES | CC(O)=C(C#N)C(=O)NC1=CC=C(C=C1)C(F)(F)F |
Nombre IUPAC | 2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide |
Peso molecular (g/mol) | 270.21 |