Learn More
Tetrahidropiran-2-ilmetilamina, 97 %, Thermo Scientific™
158.00€ - 305.00€
CAS | 683233-12-7 |
---|---|
Fórmula molecular | C6H14ClNO |
Peso molecular (g/mol) | 151.634 |
Número MDL | MFCD06738971 |
Clave InChI | NGHRATGQJXKQIQ-UHFFFAOYSA-N |
Sinónimo | oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride |
PubChem CID | 43811037 |
Nombre IUPAC | oxan-2-ilmetanamina; clorhidrato |
SMILES | C1CCOC(C1)CN.Cl |
Código de producto | Marca | Cantidad | Envase | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Envase | Precio | Cantidad y disponibilidad | ||||
10799405
|
Thermo Scientific Maybridge
CC41213CB |
250 mg | Frasco de vidrio ámbar |
158.00€
250 miligramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
10735494
|
Thermo Scientific Maybridge
CC41213DA |
1 g | Frasco de vidrio ámbar |
305.00€
1 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
683233-12-7 | |
151.634 | |
NGHRATGQJXKQIQ-UHFFFAOYSA-N | |
43811037 | |
C1CCOC(C1)CN.Cl |
C6H14ClNO | |
MFCD06738971 | |
oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride | |
oxan-2-ilmetanamina; clorhidrato |
Especificaciones
Tetrahydropyran-2-ylmethylamine hydrochloride | |
94.5 | |
95% | |
MFCD06738971 | |
NGHRATGQJXKQIQ-UHFFFAOYSA-N | |
oxan-2-ilmetanamina; clorhidrato | |
43811037 | |
95% |
683233-12-7 | |
100.0 | |
C6H14ClNO | |
oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride | |
C1CCOC(C1)CN.Cl | |
151.634 | |
151.64 |
Seguridad y manipulación
Declaración de GHS H:
Causa irritación ocular grave.
Puede irritar las vías respiratorias.
Causa irritación cutánea.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.