missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Marca: R&D Systems MAB243-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TGF-beta 3 Monoclonal specifically detects TGF-beta 3 in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Especificaciones
| TGF-beta 3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7043 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Neutralization | |
| 20724 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.1-0.3 ug/mL | |
| P10600 | |
| TGFB3 | |
| Spodoptera frugiperda, Sf 21 (baculovirus) derived recombinant human TGF-beta 3 Ala301-Ser412 (Tyr340Phe) Accession # P10600 | |
| 100 μg | |
| Primary | |
| Detects TGF-beta 3 from multiple species in direct ELISAs and Western blots. In Western blots, less than 25% cross-reactivity with recombinant human (rh) TGF-beta 1.2 and rhTGF-beta 2 is observed, and less than 2% cross-reactivity with recombinant amphibian TGF-beta 5 and recombinant human TGF-beta 1 is observed. Neutralizes the biological activity of TGF-beta 3 but not TGF-beta 1, TGF-beta 2, or TGF-beta 5. | |
| Multi-species | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido