missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Themis Antibody (701010) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB6929B
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Themis Monoclonal antibody specifically detects Themis in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Especificaciones
Themis | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
387357 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry (Intracellular Staining), CyTOF | |
701010 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
E. coli-derived recombinant human Themis, Ala2-Pro282, Accession # Q8N1K5 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Themis Antibody (701010) [Biotin], Novus Biologicals™ > 0.1 mL; Biotin
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido