Learn More
Thermo Scientific™ Thermolyne™ Premium Large Muffle Furnaces Accessories
Use these accessories with Thermolyne Premium Large Muffle Furnaces.
Marca: Thermo Scientific™ SH408X1
Detalles adicionales : Peso : 0.50000kg
Descripción
Use Thermo Scientific™ Thermolyne Premium Large Muffle Furnace Accessories to increase the utility of the Large Muffle Furnace.
Especificaciones
Estante de acero inoxidable | |
Estante | |
12,8 in |
Horno de mufla de gran tamaño Thermolyne Premium | |
8,3 in |