missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 16 publications
Marca: Novus Biologicals NB100-56571
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TLR3 Polyclonal specifically detects TLR3 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Flow (Cell Surface), Flow (Intracellular), Knockdown Validated.
Especificaciones
TLR3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
CD283, CD283 antigen, toll-like receptor 3 | |
Rabbit | |
104 kDa | |
0.1 mg | |
Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7098 | |
Human, Mouse, Pig | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, KnockDown | |
1.0 mg/mL | |
Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Patole et al (2005)), Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Pawar et al (2005)), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (Pawar et al (2006)), Flow (Intracellular) 1 ug/1 million cells, KnockDown Validated | |
O15455 | |
TLR3 | |
A mix of synthetic peptides corresponding to amino acids 135-150 (SIHKIKSNPFKNQKNL), 828-844 (CRRFKVHHAVQQAIEQN), and 876-891 (CILNWPVQKERINAFH) of mouse TLR3. | |
Protein G purified | |
RUO | |
Primary | |
Mouse and Human. Predicted to cross-react with Rat. Porcine data from customer review. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
TLR3 Antibody, Novus Biologicals™ > 0.1mg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido