missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Bioactivity, Block/Neutralize.
Specifications
Specifications
| Antígeno | TLR3 |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Neutralization |
| Clasificación | Monoclonal |
| Clon | 40C1285.6 |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1-3 ug/ml, Flow Cytometry 2-4 ug/ 1x10^6 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2 ug/ 1x10^6 cells, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) reported in scientific literature (PMID 24836676), Bioactivity reported in scientific literature (PMID 28894449), Block/Neutralize reported in scientific literature (PMID 28894449) |
| N.º de referencia del gen | O15455 |
| Alias de gen | CD283, CD283 antigen, toll-like receptor 3 |
| Símbolos de los genes | TLR3 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?