missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMOD1 Mouse anti-Human, Mouse, Rat, Clone: OTI2C2, TrueMAB™, Origene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA503170S
Detalles adicionales : Peso : 0.05000kg
Descripción
TMOD1 Monoclonal antibody specifically detects TMOD1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotEspecificaciones
TMOD1 | |
Monoclonal | |
0.86 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P28289, P49813, P70567 | |
Mouse | |
Affinity chromatography | |
RUO | |
21916, 25566, 7111 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2C2 | |
Unconjugated | |
TMOD1 | |
D9S57E; E tropomodulin 1; EGK_07519; Erythrocyte tropomodulin; ETMOD; E-Tmod; e-tropomodulin; Tmod; TMOD1; tropomodulin 1; tropomodulin-1 | |
Full length human recombinant protein of human TMOD1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |