missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA514265
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with mouse based on sequence homology.
TMPRSS2 is a protein that belongs to the serine protease family. The protein contains a type II transmembrane domain, a receptor class A domain, a scavenger receptor cysteine-rich domain and a protease domain. Serine proteases are known to be involved in many physiological and pathological processes. Its gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue. The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage.Especificaciones
TMPRSS2 | |
Polyclonal | |
Unconjugated | |
TMPRSS2 | |
D16Ertd61e; EC 3.4.219; epitheliasin; FLJ41954; MGC6821; Plasmic transmembrane protein X; PP9284; PRSS10; Serine protease 10; Tmprss2; TMPS2; Transmembrane protease serine 2; Transmembrane protease serine 2 catalytic chain; Transmembrane protease serine 2 non-catalytic chain; transmembrane protease, serine 2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
7113 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
1.8 mg/mL | |
PBS with 0.09% sodium azide | |
O15393 | |
TMPRSS2 | |
KLH conjugated synthetic peptide between 314-343 amino acids from the central region of human TMPRSS2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |