missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Rabbit, Clone: TNF/1500R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 7124-RBM13-P1
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
TNF-alpha | |
Cytoplasmic and extracellular (secreted) | |
TNF/1500R | |
Unconjugated | |
NP_000585 | |
Rabbit | |
17kDa | |
Protein A/G Purified | |
RUO | |
7124 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
TNF | |
Recombinant full-length human TNF-alpha protein | |
HeLa, HL-60, or A431 cells. Pancreas or Histiocytoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |